Invitrogen
Manufacturer Code:PA5103407
Catalog # PIPA5103407
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human DNAJB5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110058L06Rik AI462558 DnaJ (Hsp40) homolog subfamily B member 5 DnaJ heat shock protein family (Hsp40) member B5 dnaJ homolog subfamily B member 5 DNAJB5 heat shock cognate 40 heat shock protein cognate 40 Heat shock protein Hsp40-2 heat shock protein Hsp40-3 Hsc40 Hsp40-3; DnaJ (Hsp40) homolog, subfamily B, member 5; DnaJ homolog subfamily B member 5; heat shock cognate 40; Heat shock protein cognate 40; Heat shock protein Hsp40-2; Heat shock protein Hsp40-3; Hsc40
Gene Aliases: 1110058L06Rik; AI462558; DNAJB5; HSC40; Hsp40-3
UniProt ID: (Human) O75953, (Mouse) O89114
Entrez Gene ID: (Human) 25822, (Mouse) 56323
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.