R&D Systems
Manufacturer Code:AB4315
Catalog # AB4315
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human DLL3 Ala27-Pro488 Accession # Q9NYJ7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: delta (Drosophila)-like 3 Delta3 delta-like 3 (Drosophila) delta-like protein 3 Drosophila Delta homolog 3 Pudgy SCDO1 SCDO1delta3; delta-like 3; Delta-like protein 3; Delta3; Drosophila Delta homolog 3
Gene Aliases: DLL3; SCDO1
UniProt ID: (Human) Q9NYJ7
Entrez Gene ID: (Human) 10683
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.