Invitrogen
Manufacturer Code:PA583742
Catalog # PIPA583742
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human DLD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: diaphorase; Dihydrolipoamide dehydrogenase; Dihydrolipoyl dehydrogenase, mitochondrial; DLDH GCSL LAD; E3 component of pyruvate dehydrogenase complex, 2-oxo-glutarate complex, branched chain keto acid dehydrogenase complex; Glycine cleavage system L protein; glycine cleavage system protein L; lipoamide dehydrogenase; lipoamide reductase; lipoyl dehydrogenase
Gene Aliases: DLD; DLDD; DLDH; E3; GCSL; LAD; PHE3
UniProt ID: (Human) P09622
Entrez Gene ID: (Human) 1738
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.