Invitrogen
Manufacturer Code:PA5116502
Catalog # PIPA5116502
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-189 of human PARK7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cap1 Contraception-associated protein 1 CTA-215D11.1 DJ1 DJ-1 DJ-1 protein early onset 7 epididymis secretory sperm binding protein Li 67p fertility protein SP22 FLJ27376 FLJ34360 FLJ92274 HEL-S-67p hypothetical protein LOC449674 maillard deglycase Oncogene DJ1 OTTHUMP00000001350 OTTHUMP00000001351 park7 PARK7 GENE PARK7/DJ1 parkinson disease (autosomal recessive early onset) 7 Parkinson disease 7 Parkinson disease protein 7 Parkinson disease protein 7 homolog Parkinson disease protein 7 homolog protein/nucleic acid deglycase DJ-1 parkinson protein 7 Parkinsonism associated deglycase parkinsonism-associated deglycase Protein CAP1 protein deglycase DJ-1 protein deglycase DJ-1zDJ-1 Protein DJ-1 protein DJ-1-like protein protein DJ-1zDJ-1 Protein/nucleic acid deglycase DJ-1 RNA-binding protein regulatory subunit SP22 zDJ-1 zgc:103725; DJ-1; epididymis secretory sperm binding protein Li 67p; Maillard deglycase; Oncogene DJ1; Parkinson disease (autosomal recessive, early onset) 7; Parkinson disease protein 7; Parkinson disease protein 7 homolog; parkinson protein 7; Parkinsonism-associated deglycase; protein deglycase DJ-1; Protein DJ-1; Protein/nucleic acid deglycase DJ-1; RNA-binding protein regulatory subunit
Gene Aliases: DJ-1; DJ1; HEL-S-67p; PARK7
UniProt ID: (Human) Q99497, (Mouse) Q99LX0
Entrez Gene ID: (Human) 11315, (Mouse) 57320
Molecular Function: RNA binding protein cysteine protease hydrolase nucleic acid binding protease transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.