Invitrogen
Manufacturer Code:PA579152
Catalog # PIPA579152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived human DHFR recombit protein (Position: V2-D187). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Dihydrofolate reductase; Dihydrofolate reductase 1 (active); Dihydrofolate reductase DHFR
Gene Aliases: 8430436I03Rik; AA607882; AI662710; AW555094; DHFR; Dhfr1; DHFRP1; DYR
UniProt ID: (Human) P00374, (Mouse) P00375, (Rat) Q920D2
Entrez Gene ID: (Human) 1719, (Mouse) 13361, (Rat) 24312
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.