Invitrogen
Manufacturer Code:PA597912
Catalog # PIPA597912
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Keratin type II cytoskeletal 6B protein (1-280AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CK-6B; cytokeratin 6B; Cytokeratin-6B; K6B; keratin 6B, type II; keratin, epidermal, type II, K6B; Keratin, type II cytoskeletal 6B; Keratin-6B; keratin-like 1 (a type II keratin sequence); Type-II keratin Kb10
Gene Aliases: CK-6B; CK6B; K6B; KRT6B; KRTL1; PC2; PC4
UniProt ID: (Human) P04259
Entrez Gene ID: (Human) 3854
Molecular Function: cytoskeletal protein intermediate filament structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.