Novus Biologicals
Manufacturer Code:NBP140974PCP
Catalog # NBP140974P
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CS9 |
Immunogen | Cytochrome B245 solubilised in octyl-beta-gluycopyranoside |
Conjugate | PerCP |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: cytochrome b light chain; cytochrome b light chain Cytochrome b(558) alpha chain cytochrome b(558) alpha-subunit cytochrome b alpha polypeptide cytochrome b-245 light chain cytochrome b-245 alpha polypeptide Cytochrome b558 subunit alpha flavocytochrome b-558 alpha polypeptide Neutrophil cytochrome b 22 kDa polypeptide p22 phagocyte B-cytochrome p22phox p22-phox Superoxide-generating NADPH oxidase light chain subunit; Cytochrome b(558) alpha chain; cytochrome b(558) alpha-subunit; cytochrome b, alpha polypeptide; Cytochrome b-245 light chain; cytochrome b-245, alpha polypeptide; Cytochrome b558 subunit alpha; flavocytochrome b-558 alpha polypeptide; Neutrophil cytochrome b 22 kDa polypeptide; p22 phagocyte B-cytochrome; p22-phox; p22phox; Superoxide-generating NADPH oxidase light chain subunit
Gene Aliases: CYBA; p22-PHOX
UniProt ID: (Human) P13498
Entrez Gene ID: (Human) 1535
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.