Invitrogen
Manufacturer Code:PA518044
Catalog # PIPA518044
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
peptide-ELISA (pep-ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (PWLQHFEVPVLRCH) corresponding to the C-terminus amino acids of CST7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: CMAP; CST7 cystatin F (leukocystatin) CMAP cystatin 7 leukocystatin cystatin-like metastasis-associated protein cystatin F dJ568C11.1 (cystatin F (leukocystatin)); cystatin 7; Cystatin-7; Cystatin-F; Cystatin-like metastasis-associated protein; Leukocystatin
Gene Aliases: Cmap; Cst7
UniProt ID: (Mouse) O89098
Entrez Gene ID: (Mouse) 13011
Molecular Function: cysteine protease inhibitor enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.