Novus Biologicals
Manufacturer Code:NBP298950AF350
Catalog # NB082945
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Cystatin F (NP_003641.2 Met 1-His 145). |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMAP; CMAP cystatin F (leukocystatin) Cystatin-7 cystatin-F Cystatin-like metastasis-associated protein leukocystatin; cystatin F (leukocystatin); Cystatin-7; Cystatin-F; Cystatin-like metastasis-associated protein; Leukocystatin
Gene Aliases: CMAP; CST7
UniProt ID: (Human) O76096
Entrez Gene ID: (Human) 8530
Molecular Function: cysteine protease inhibitor enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.