Novus Biologicals
Manufacturer Code:NBP289501100UL
Catalog # NB020485
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 33 |
Immunogen | Recombinant Human Cystatin C protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: ARMD11 bA218C14.4 (cystatin C) cystatin 3 cystatin C cystatin C (amyloid angiopathy and cerebral hemorrhage) cystatin-3 cystatin-C Gamma-trace MGC117328 Neuroendocrine basic polypeptide post-gamma-globulin; bA218C14.4 (cystatin C); cystatin 3; Cystatin-3; Cystatin-C; epididymis secretory protein Li 2; Gamma-trace; Neuroendocrine basic polypeptide; Post-gamma-globulin
Gene Aliases: ARMD11; CST3; HEL-S-2
UniProt ID: (Human) P01034
Entrez Gene ID: (Human) 1471
Molecular Function: cysteine protease inhibitor enzyme modulator protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.