Novus Biologicals
Manufacturer Code:NB1002612AF532
Catalog # N102612A532
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | X121.10 |
Immunogen | Xenopus laevis Cyclin B2 |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: cyclin B2 G2/mitotic-specific cyclin-B2 HsT17299; G2/mitotic-specific cyclin-B2
Gene Aliases: CCNB2; HsT17299
UniProt ID: (Human) O95067
Entrez Gene ID: (Human) 9133
Molecular Function: enzyme modulator kinase activator kinase modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.