Abnova
Manufacturer Code:PAB15731
Catalog # 89-117-796
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 138 mouse Csde1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Csde1.
For Research Use Only
Protein Aliases: AA960392; BC016898; Cold shock domain-containing protein E1; D3Jfr1; MGC19174; mKIAA0885; NRAS-related; unr; upstream of NRAS
Gene Aliases: AA960392; BC016898; Csde1; D3Jfr1; mKIAA0885; unr
UniProt ID: (Mouse) Q91W50
Entrez Gene ID: (Mouse) 229663
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.