Novus Biologicals
Manufacturer Code:NBP237568
Catalog # NBP237568
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3G11E8 |
Immunogen | Purified recombinant fragment of human CRK expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Adapter molecule crk; adapter molecule crk avian sarcoma virus CT10 (v-crk) oncogene homolog CRKII p38 Proto-oncogene c-Crk v-crk avian sarcoma virus CT10 oncogene homolog v-crk sarcoma virus CT10 oncogene homolog (avian); p38; Proto-oncogene c-Crk; v-crk avian sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene-like protein
Gene Aliases: CRK; CRKII; p38
UniProt ID: (Human) P46108
Entrez Gene ID: (Human) 1398
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.