Novus Biologicals
Manufacturer Code:NBP298381F
Catalog # NB082336
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Contactin-2/TAG1 (Q61330 Gln31-Glu1013). |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Axonal glycoprotein TAG-1; Axonal glycoprotein TAG-1 Axonin-1 axonin-1 cell adhesion molecule AXT contactin 2 (axonal) contactin 2 (transiently expressed) contactin-2 DKFZp781D102 FLJ37193 MGC157722 TAG1 TAG-1 TAX TAX-1 TAX1FLJ42746 Transient axonal glycoprotein 1 transiently-expressed axonal glycoprotein; Axonin-1; axonin-1 cell adhesion molecule; contactin 2 (axonal); contactin 2 (transiently expressed); Contactin-2; TAX-1; Transient axonal glycoprotein 1; transiently-expressed axonal glycoprotein
Gene Aliases: AXT; CNTN2; FAME5; TAG-1; TAG1; TAX; TAX1
UniProt ID: (Human) Q02246
Entrez Gene ID: (Human) 6900
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.