Invitrogen
Manufacturer Code:PA587083
Catalog # PIPA587083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human CNTN2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Axonal glycoprotein TAG-1; axonin; Axonin-1; axonin-1 cell adhesion molecule; contactin 2 (axonal); contactin 2 (transiently expressed); Contactin 2 Contactin2 Axonal glycoprotein TAG-1 Axonin-1 Axonin1 Axonin 1 Transient axonal glycoprotein 1 TAX-1 CNTN2 AXT TAG1 TAX1; Contactin-2; TAX-1; Transient axonal glycoprotein 1; transiently-expressed axonal glycoprotein
Gene Aliases: AXT; CNTN2; D130012K04Rik; FAME5; TAG-1; TAG-564; TAG1; TAX; TAX1
UniProt ID: (Human) Q02246, (Mouse) Q61330, (Rat) P22063
Entrez Gene ID: (Human) 6900, (Mouse) 21367, (Rat) 25356
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.