R&D Systems
Manufacturer Code:MAB4920
Catalog # MAB4920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 544623 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human Carboxylesterase1/CES-1 His19-Glu563 Accession # P23141 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACAT; ACAT acyl coenzyme A:cholesterol acyltransferase Acyl-coenzyme A:cholesterol acyltransferase Brain carboxylesterase hBr1 carboxylesterase 1 carboxylesterase 1 (monocyte/macrophage serine esterase 1) carboxylesterase 2 (liver) CEH Ces-1 CES1 CES1A2 CES2EC 3.1.1.1 cholesteryl ester hydrolase Cocaine carboxylesterase EC 3.1.1 egasyn HMSE1 HMSECES1A1 human monocyte/macrophage serine esterase 1 liver carboxylesterase 1 MGC117365 Monocyte/macrophage serine esterase PCE-1 REH Retinyl ester hydrolase Serine esterase 1 Ses-1 SES1 SES1TGH TGH Triacylglycerol hydrolase; acyl coenzyme A:cholesterol acyltransferase; Acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; Carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 2 (liver); CE-1; CEH; Cholesteryl ester hydrolase; Cocaine carboxylesterase; Egasyn; HMSE; human monocyte/macrophage serine esterase 1; Liver carboxylesterase 1; Methylumbelliferyl-acetate deacetylase 1; Monocyte/macrophage serine esterase; REH; Retinyl ester hydrolase; Serine esterase 1; TGH; Triacylglycerol hydrolase
Gene Aliases: ACAT; CE-1; CEH; CES1; CES2; hCE-1; HMSE; HMSE1; PCE-1; REH; SES1; TGH
UniProt ID: (Human) P23141
Entrez Gene ID: (Human) 1066
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.