Invitrogen
Manufacturer Code:PA577295
Catalog # PIPA577295
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)PSSPERAPGREGPYGRE corresponding to amino acid residues 865-881 of rat CaV 2.1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha1A Cacna1a; BI; brain calcium channel 1; Brain calcium channel I; brain class A; calcium channel alpha 1A; calcium channel, L type, alpha-1 polypeptide; Calcium channel, L type, alpha-1 polypeptide isoform 4; Calcium channel, L type, alpha-1 polypeptide, isoform 4; calcium channel, voltage-dependent, alpha 1A subunit; calcium channel, voltage-dependent, P/Q type, alpha 1A subunit; fetal brain Ca2+ voltage-gated channel alpha1A pore-forming subunit; leaner; Rat brain class A; RBA-I; rocker; tottering; Voltage-dependent P/Q-type calcium channel subunit alpha-1A; Voltage-gated calcium channel subunit alpha Cav2.1
Gene Aliases: alpha1A; APCA; BccA1; BI; Caca1a; CACH4; CACN3; CACNA1A; CACNL1A4; CAV2.1; Ccha1a; EA2; FHM; HPCA; la; MHP; MHP1; nmf352; rbA-1; rkr; SCA6; tg
UniProt ID: (Human) O00555, (Mouse) P97445, (Rat) P54282
Entrez Gene ID: (Human) 773, (Mouse) 12286, (Rat) 25398
Molecular Function: calcium channel ion channel sodium channel transporter voltage-gated calcium channel voltage-gated ion channel voltage-gated sodium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.