Novus Biologicals
Manufacturer Code:NBP250214IR
Catalog # NBP250214IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | M29-P3B10 |
Immunogen | Peptide sequence - KREELGLEGQ (amino acids 495 - 504) |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BCD CYP4AH1 cytochrome P450 4V2 cytochrome P450 family 4 subfamily V polypeptide 2 EC 1.14 EC 1.14.14.1 FLJ18432 MGC43534; Cytochrome P450 4V2; cytochrome P450, family 4, subfamily V, polypeptide 2; Docosahexaenoic acid omega-hydroxylase CYP4V2; Long-chain fatty acid omega-monooxygenase
Gene Aliases: BCD; CYP4AH1; CYP4V2
UniProt ID: (Human) Q6ZWL3
Entrez Gene ID: (Human) 285440
Molecular Function: oxidoreductase oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.