Invitrogen
Manufacturer Code:PA539581
Catalog # PIPA539581
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide derived from the C-terminal region of human Cytochrome P450 11B1/2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ALDOS; Aldosterone synthase; Aldosterone-synthesizing enzyme; Corticosterone 18-monooxygenase, CYP11B2; CPN1 CYP11B FHI P450C11 CYPXIB1 cytochrome P-450c11 cytochrome P450 11B1 mitochondrial cytochrome P450 subfamily XIB (steroid 11-beta-hydroxylase) polypeptide 1 cytochrome P450C11 cytochrome p450 XIB1 steroid 11-beta-hydroxylase steroid 11-beta-monooxygenase; CYPXIB1; CYPXIB2; Cytochrome P-450Aldo; Cytochrome P-450c11; Cytochrome P-450C18; Cytochrome P450 11B1, mitochondrial; Cytochrome P450 11B2, mitochondrial; cytochrome p450 XIB1; cytochrome P450, family 11, subfamily B, polypeptide 1; cytochrome P450, family 11, subfamily B, polypeptide 2; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1; cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 2; Cytochrome P450C11; mitochondrial cytochrome P450, family 11, subfamily B, polypeptide 2; steroid 11-beta-hydroxylase; Steroid 11-beta-hydroxylase, CYP11B1; Steroid 11-beta-hydroxylase, CYP11B2; steroid 11-beta-monooxygenase; steroid 11-beta/18-hydroxylase; Steroid 18-hydroxylase; steroid 18-hydroxylase, aldosterone synthase, P450C18, P450aldo
Gene Aliases: ALDOS; CPN1; CPN2; CYP11B; CYP11B1; CYP11B2; CYP11BL; CYPXIB2; FHI; P-450C18; P450aldo; P450C11; P450C18; S11BH
UniProt ID: (Human) P15538, (Human) P19099
Entrez Gene ID: (Human) 1584, (Human) 1585
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.