Invitrogen
Manufacturer Code:PA580029
Catalog # PIPA580029
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human SLC31A1/CTR1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: copper transport 1 homolog; Copper transporter 1; Copper uptake transporter 1; CTR1; hCTR1; High affinity copper uptake protein 1; High affinity copper uptake protein 1 Copper transporter 1 hCTR1 Solute carrier family 31 member 1 SLC31A1 COPT1 CTR1; Liver regeneration-related protein LRRGT00200; rCTR1; solute carrier family 13 (sodium/sulphate symporters), member 1; solute carrier family 31 (copper transporter), member 1; solute carrier family 31 (copper transporters), member 1; Solute carrier family 31 member 1
Gene Aliases: 4930445G01Rik; AI787263; AU016967; COPT1; CTR1; LRRGT00200; SLC31A1
UniProt ID: (Human) O15431, (Rat) Q9JK41, (Mouse) Q8K211
Entrez Gene ID: (Human) 1317, (Rat) 171135, (Mouse) 20529
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.