Novus Biologicals
Manufacturer Code:NBP171913
Catalog # NBP171913
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 50 and 100 of human Upstream of NRAS using the numbering given in entry NP_001007554.1 (GeneID 7812). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cold shock domain containing E1 RNA-binding cold shock domain-containing protein E1 D1S155EUNRRP5-1000E10.3 DKFZp779B0247 DKFZp779J1455 FLJ26882 KIAA0885 N-ras upstream gene protein NRAS-related NRU Protein UNR upstream of NRAS; cold shock domain containing E1, RNA binding; cold shock domain containing E1, RNA-binding; Cold shock domain-containing protein E1; N-ras upstream gene protein; NRAS-related; Protein UNR; upstream of NRAS
Gene Aliases: CSDE1; D1S155E; KIAA0885; NRU; UNR
UniProt ID: (Human) O75534
Entrez Gene ID: (Human) 7812
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.