Invitrogen
Manufacturer Code:PA543759
Catalog # PIPA543759
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human CRYM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CDK108; Crystallin mu DFNA40 THBP; crystallin, mu; Ketimine reductase mu-crystallin; mu-crystallin homolog; Mu-crystallin-like protein; NADP-regulated thyroid-hormone binding protein; NADP-regulated thyroid-hormone-binding protein; thiomorpholine-carboxylate dehydrogenase
Gene Aliases: CRYM; DFNA40; THBP
UniProt ID: (Human) Q14894, (Bovine) Q2KHX6, (Rat) Q9QYU4, (Mouse) O54983
Entrez Gene ID: (Rabbit) 100352494, (Human) 1428, (Dog) 479818, (Goat) 100861024, (Bovine) 505167, (Horse) 100057895, (Rat) 117024, (Guinea pig) 100717691, (Mouse) 12971
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.