Novus Biologicals
Manufacturer Code:NBP151952
Catalog # NBP151952
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-APLKPLPPSAKDPA corresponding to C-Terminus according to NP_690902.1 NP_003906.2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Chromobindin 17; Copine I; copine ICPN1COPN1 copine-1 MGC1142; Copine-1
Gene Aliases: COPN1; CPN1; CPNE1
UniProt ID: (Human) Q99829
Entrez Gene ID: (Human) 8904
Molecular Function: membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.