Proteintech
Manufacturer Code:169491AP150UL
Catalog # 50-173-1286
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CPLX3 Fusion Protein Ag10571 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: complexin 3 Complexin III CPLX3 CPX III CPXIII Nbla11589; Complexin III; Complexin-3; CPX III; lectin, mannose-binding, 1 like
Gene Aliases: BC016632; CPLX3; CPX-III; CPXIII; ERGIC-53L; ERGL; Lamn1l; Lman1l; Nbla11589; RGD1560104
UniProt ID: (Human) Q8WVH0, (Mouse) Q8R1B5
Entrez Gene ID: (Human) 594855, (Mouse) 235415, (Rat) 501005
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.