Invitrogen
Manufacturer Code:PA526419
Catalog # PIPA526419
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 33-60 amino acids from the central region of human CPLX1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 921-S; Complexin I; Complexin-1; CPX I; CPX-I CPX1 CPX I complexin I complexin-1 synaphin 2 synaphin-2; synaphin 2; Synaphin-2
Gene Aliases: 921-S; CPLX1; CPX-I; CPX1
UniProt ID: (Human) O14810, (Mouse) P63040
Entrez Gene ID: (Human) 10815, (Mouse) 12889
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.