Invitrogen
Manufacturer Code:PA5101305
Catalog # PIPA5101305
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human COX17 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI037035 Cox17 COX17 cytochrome c oxidase assembly homolog COX17 cytochrome c oxidase copper chaperone COX17 homolog cytochrome c oxidase assembly protein COX17 cytochrome c oxidase copper chaperone Cox17a cytochrome c oxidase 17 copper chaperone cytochrome c oxidase assembly homolog 17 cytochrome c oxidase assembly protein 17 cytochrome c oxidase copper chaperone cytochrome c oxidase subunit XVII assembly protein homolog cytochrome c oxidase subunit XVII assembly protein homolog human homolog of yeast mitochondrial copper recruitment; COX17 cytochrome c oxidase assembly homolog; COX17 cytochrome c oxidase copper chaperone; COX17 homolog, cytochrome c oxidase assembly protein; cytochrome c oxidase 17 copper chaperone; cytochrome c oxidase assembly homolog 17; Cytochrome c oxidase copper chaperone; cytochrome c oxidase subunit XVII assembly protein homolog; cytochrome c oxidase, subunit XVII assembly protein homolog; human homolog of yeast mitochondrial copper recruitment
Gene Aliases: AI037035; COX17; Cox17a
UniProt ID: (Human) Q14061, (Mouse) P56394
Entrez Gene ID: (Human) 10063, (Rat) 89786, (Mouse) 12856
Molecular Function: transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.