Novus Biologicals
Manufacturer Code:NBP272224IR
Catalog # NBP272224IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3B3 |
Immunogen | Full length recombinant protein of human COASY (NP_079509) produced in HEK293T cells. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Bifunctional coenzyme A synthase; bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase; bifunctional phosphopantetheine adenylyl transferase/dephospho CoA kinase CoA synthaseUKR1 CoASY Coenzyme A synthase DPCK FLJ35179 NBPbifunctional coenzyme A synthase nucleotide binding protein phosphopantetheine adenylyltransferase/ dephosphocoenzyme A kinase POV-2 PPAT; CoA synthase; Dephospho-CoA kinase; Dephospho-CoA pyrophosphorylase; Dephosphocoenzyme A kinase; DPCK; DPCOAK; NBP; nucleotide binding protein; Pantetheine-phosphate adenylyltransferase; Phosphopantetheine adenylyltransferase; phosphopantetheine adenylyltransferase / dephosphocoenzyme A kinase; POV-2; PPAT
Gene Aliases: COASY; DPCK; NBIA6; NBP; pOV-2; PPAT; PSEC0106; UKR1
UniProt ID: (Human) Q13057
Entrez Gene ID: (Human) 80347
Molecular Function: kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.