Proteintech
Manufacturer Code:661731IG150UL
Catalog # 50-173-6607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G8G11 |
Immunogen | CNPY2 MSAP Fusion Protein Ag6363 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: canopy 2 homolog; canopy 2 homolog (zebrafish) CNPY2 CNPY2MSAP HP10390 MSAP Protein canopy homolog 2 TMEM4 Transmembrane protein 4 ZSIG9; MIR-interacting saposin-like protein; Protein canopy homolog 2; Putative secreted protein ZSIG9; Transmembrane protein 4
Gene Aliases: 5330432A10Rik; AW229003; CNPY2; D10Bwg1546e; HP10390; MSAP; TMEM4; UNQ1943/PRO4426; ZSIG9
UniProt ID: (Human) Q9Y2B0, (Mouse) Q9QXT0
Entrez Gene ID: (Human) 10330, (Mouse) 56530, (Rat) 685814
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.