Novus Biologicals
Manufacturer Code:NBP186564
Catalog # NBP186564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:RLLNYLLDNLSGTAKRITTEQPPPRSWIMLQEPDRTRPTALFSVMCYN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: carbon catabolite repression 4 protein; carbon catabolite repression 4 protein Carbon catabolite repressor protein 4 homolog CCR4 carbon catabolite repression 4-like CCR4CCR4-NOT transcription complex subunit 6 CCR4-NOT transcription complex subunit 6 Cytoplasmic deadenylase EC 3.1.13.4 KIAA1194EC 3.1; carbon catabolite repressor protein 4 homolog; CCR4-NOT transcription complex, subunit 6; cytoplasmic deadenylase
Gene Aliases: CCR4; Ccr4a
Entrez Gene ID: (Human) 57472
Molecular Function: RNA binding protein exoribonuclease nuclease nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.