Novus Biologicals
Manufacturer Code:NBP179946
Catalog # NBP179946
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N terminal of human CHMThe immunogen for this antibody is CHM. Peptide sequence LHVDSRSYYGGNWASFSFSGLLSWLKEYQENSDIVSDSPVWQDQILENEE. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: choroideraemia protein choroideremia (Rab escort protein 1) Rab geranylgeranyltransferase component A rab proteins geranylgeranyltransferase component A 1 REP1; choroideremia (Rab escort protein 1); Choroideremia protein; Rab escort protein 1; Rab proteins geranylgeranyltransferase component A 1; REP-1; TCD protein
Gene Aliases: CHM; DXS540; GGTA; HSD-32; REP-1; REP1; TCD
UniProt ID: (Human) P24386
Entrez Gene ID: (Human) 1121
Molecular Function: G-protein modulator acyltransferase enzyme modulator transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.