Proteintech
Manufacturer Code:121691AP150UL
Catalog # 50-172-7765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHFR Fusion Protein Ag2811 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Checkpoint with forkhead and RING finger domains protein; CHFR RING finger protein 196 RNF116 RNF196; E3 ubiquitin-protein ligase CHFR; RING finger protein 196; RING-type E3 ubiquitin transferase CHFR
Gene Aliases: 5730484M20Rik; C230082M18; CHFR; RNF116; RNF196
UniProt ID: (Human) Q96EP1, (Mouse) Q810L3
Entrez Gene ID: (Human) 55743, (Rat) 288734, (Mouse) 231600
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.