Abnova
Manufacturer Code:MAB1069
Catalog # 89-108-890
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 47A1 |
Immunogen | Recombinant protein corresponding to amino acids 1-271 of human CEBPB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant CEBPB.
For Research Use Only
Protein Aliases: C/EBP beta; C/EBP-beta; CCAAT/enhancer binding protein (C/EBP), beta; CCAAT/enhancer-binding protein beta; CRP2; IL6DBP; interleukin 6-dependent DNA-binding protein; LAP; LIP; Liver activator protein; Liver-enriched inhibitory protein; MGC32080; NF-IL6; Nuclear factor NF-IL6; nuclear factor of interleukin 6; TCF-5; TCF5; Transcription factor 5; transcription factor C/EBP beta
Gene Aliases: C/EBP-beta; CEBPB; IL6DBP; NF-IL6; PP9092; TCF5
UniProt ID: (Human) P17676
Entrez Gene ID: (Human) 1051
Molecular Function: nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.