Invitrogen
Manufacturer Code:PA587248
Catalog # PIPA587248
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human CEACAM1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: antigen CD66; ATP-dependent taurocolate-carrier protein; BGP-1; Biliary glycoprotein 1; Biliary glycoprotein D; bone gamma-carboxyglutamic acid (Gla) protein; C-CAM 105; carcinoembryonic antigen 1; carcinoembryonic antigen 7; Carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); Carcinoembryonic antigen-related cell adhesion molecule 1 Biliary glycoprotein 1 BGP-1 CD66a CEACAM1 BGP BGP1; CD66a; CD66a antigen; CEA-related cell adhesion molecule 1; CEA-related cell adhesion molecule 1 (bone gamma-carboxyglutamic acid (Gla) protein) (osteocalcin); Cell-CAM 105; Ecto-ATPase; GP110; hepatitis virus (MHV-4) susceptibility; hepatitis virus receptor; MHV-R; MHVR1; Murine hepatitis virus receptor; osteocalcin; pp120; pp120/ecto-ATPase
Gene Aliases: bb-1; BGP; BGP1; BGPI; BGPR; C-CAM; Cc1; Ccam1; CD66a; Cea-1; Cea-7; Cea1; Cea7; CEACAM1; Hv-2; Hv2; mCEA1; Mhv-1; MHVR; MHVR1; mmCGM1; mmCGM1a; mmCGM2
UniProt ID: (Human) P13688, (Rat) P16573, (Mouse) P31809
Entrez Gene ID: (Human) 634, (Rat) 81613, (Mouse) 26365
Molecular Function: cell adhesion molecule cytokine receptor defense/immunity protein immunoglobulin receptor superfamily kinase membrane-bound signaling molecule protein kinase receptor signaling molecule transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.