Novus Biologicals
Manufacturer Code:NBP237330
Catalog # NBP237330
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7E1B11 |
Immunogen | Purified recombinant fragment of human THY1 (AA: 17-132) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CD90; CD90 CD90 antigen CDw90 FLJ33325 Thy-1 antigen Thy-1 cell surface antigen thy-1 membrane glycoprotein Thy-1 T-cell antigen; CDw90; Thy-1 antigen; Thy-1 membrane glycoprotein; Thy-1 T-cell antigen
Gene Aliases: CD90; CDw90; THY1
UniProt ID: (Human) P04216
Entrez Gene ID: (Human) 7070
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.