Proteintech
Manufacturer Code:183301AP150UL
Catalog # 50-173-2117
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | CD24 Fusion Protein Ag11679 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CD24; CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24 CD24 molecule CD24A Signal transducer CD24; cluster of differentiation 24; heat stable antigen; HSA; Ly-52; Lymphocyte antigen 52; M1/69-J11D heat stable antigen; Nectadrin; R13-Ag; Signal transducer CD24; Small cell lung carcinoma cluster 4 antigen; X62 heat stable antigen
Gene Aliases: CD24; CD24A; HSA; Ly-52; nectadrin
UniProt ID: (Human) P25063, (Mouse) P24807
Entrez Gene ID: (Human) 100133941, (Mouse) 12484
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.