Invitrogen
Manufacturer Code:46122282
Catalog # 50-245-876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | TM-b1 (TM-beta1) |
Conjugate | PerCP-eFluor 710 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD122; CD122 IL15RB P70-75 CD122 antigen IL-2 receptor subunit beta IL-2R subunit beta IL-2RB high affinity IL-2 receptor beta subunit high affinity IL-2 receptor subunit beta interleukin 15 receptor beta interleukin-2 receptor subunit beta p75; High affinity IL-2 receptor subunit beta; IL-15 receptor beta chain; IL-2 receptor beta chain; IL-2 receptor subunit beta; IL-2/15 receptor-beta; IL-2R subunit beta; IL-2RB; Interleukin-2 receptor subunit beta; p70-75
Gene Aliases: CD122; IL-15Rbeta; Il-2/15Rbeta; Il-2Rbeta; IL15Rbeta; Il2rb; p70
UniProt ID: (Mouse) P16297
Entrez Gene ID: (Mouse) 16185
Molecular Function: cytokine receptor receptor type I cytokine receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.