Novus Biologicals
Manufacturer Code:NBP292903100UL
Catalog # NB027666
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human CCT3 (NP_005989.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCT-gamma; Cctg chaperonin containing TCP1 subunit 3 (gamma) hTRiC5 TCP1 (t-complex-1) ring complex polypeptide 5 TRIC5gamma subunit; chaperonin containing TCP1, subunit 3 (gamma); hTRiC5; T-complex protein 1 subunit gamma; T-complex protein 1, gamma subunit; TCP-1-gamma; TCP1 (t-complex-1) ring complex, polypeptide 5
Gene Aliases: CCT-gamma; CCT3; CCTG; PIG48; TCP-1-gamma; TRIC5
UniProt ID: (Human) P49368
Entrez Gene ID: (Human) 7203
Molecular Function: chaperone chaperonin
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.