R&D Systems
Manufacturer Code:FAB151B
Catalog # FAB151B
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 48607 |
Immunogen | NS0 mouse myeloma cell line transfected with human CCR2 Met1-Leu360 Accession # NP_001116868 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C chemokine receptor type 2; C-C chemokine receptor type 2 C-C CKR-2 CC-CKR-2 CC-CKR-2CKR2B CCR2 CCR-2 CCR2A CCR2B CD192 CD192 antigen chemokine (C-C motif) receptor 2 CKR2 CKR2A CMKBR2 CMKBR2MGC111760 FLJ78302 MCP-1 receptor MCP-1-R MCP-1-RMGC103828 MGC168006 Monocyte chemoattractant protein 1 receptor monocyte chemotactic protein 1 receptor; C-C CKR-2; CD192; chemokine (C-C motif) receptor 2; MCP-1 receptor; MCP-1-R; Monocyte chemoattractant protein 1 receptor; monocyte chemotactic protein 1 receptor
Gene Aliases: CC-CKR-2; CCR-2; CCR2; CCR2A; CCR2B; CD192; CKR2; CKR2A; CKR2B; CMKBR2; MCP-1-R
UniProt ID: (Human) P41597
Entrez Gene ID: (Human) 729230
Molecular Function: G-protein coupled receptor cell adhesion molecule cytokine receptor defense/immunity protein hydrolase immunoglobulin receptor superfamily immunoglobulin superfamily cell adhesion molecule phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.