Invitrogen
Manufacturer Code:PA532074
Catalog # PIPA532074
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 237 of Human CBR4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-ketoacyl-[acyl-carrier-protein] reductase beta subunit; 3-oxoacyl-[acyl-carrier-protein] reductase; carbonic reductase 4; Carbonyl reductase family member 4; KAR beta subunit; Quinone reductase CBR4; SDR45C1 3-oxoacyl-&lsqbacyl-carrier-protein] reductase carbonic reductase 4 carbonyl reductase family member 4 quinone reductase CBR4 short chain dehydrogenase/reductase family 45C member 1; Short chain dehydrogenase/reductase family 45C member 1; short chain dehydrogenase/reductase family 45C, member 1
Gene Aliases: CBR4; SDR45C1
UniProt ID: (Human) Q8N4T8
Entrez Gene ID: (Human) 84869
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.