Abnova
Manufacturer Code:MAB3491
Catalog # 89-111-444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3H2 |
Immunogen | Recombinant protein corresponding to mouse Bmp4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C on dry atmosphere. After reconstitution with distilled water,store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant Bmp4.
For Research Use Only
Protein Aliases: BMP-2B; BMP-4; Bmp2b; Bmp2b-1; Bmp2b1; Bone morphogenetic protein 2B; Bone morphogenetic protein 4
Gene Aliases: Bmp-4; Bmp2b; Bmp2b-1; Bmp2b1; Bmp4; Dvr-4
UniProt ID: (Mouse) P21275
Entrez Gene ID: (Mouse) 12159
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.