Invitrogen
Manufacturer Code:PA518787
Catalog # PIPA518787
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RNYDYYASRVPESIK) corresponding to the C-terminus amino acids of BPNT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: 3'(2'),5'-bisphosphate nucleotidase 1; bisphosphate 3&prime-nucleotidase 3&prime(2&prime) 5&prime-bisphosphate nucleotidase 1 PIP BPntase PAP-inositol-14-phosphatase; Bisphosphate 3'-nucleotidase 1; BPntase; epididymis luminal protein 20; PAP-inositol 1,4-phosphatase; PAP-inositol-1,4-phosphatase; PIP; testicular tissue protein Li 29
Gene Aliases: BPNT1; HEL20; PIP
UniProt ID: (Human) O95861
Entrez Gene ID: (Human) 10380
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.