Abnova
Manufacturer Code:PAB27736
Catalog # 89-129-253
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 20 amino acid at internal region of human BEST3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of BEST3.
For Research Use Only
Protein Aliases: Bestrophin-3; MGC13168; MGC40411; vitelliform macular dystrophy 2-like 3; Vitelliform macular dystrophy 2-like protein 3; VMD2L3
Gene Aliases: BEST3; VMD2L3
UniProt ID: (Human) Q8N1M1
Entrez Gene ID: (Human) 144453
Molecular Function: anion channel ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.