Invitrogen
Manufacturer Code:MA535573
Catalog # PIMA535573
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1121 |
Immunogen | A synthesized peptide derived from human BDKRB2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: B(2) B2 B2 bradykinin receptor B2BKR B2BRA B2R Bdkrb2 BK2 BK-2 BK2R BK-2R BK-2Rs BKR2 BRB2 DKFZp686O088; B2 bradykinin receptor; B2R; BK-2 receptor; kinin B2
Gene Aliases: B(2); B2; B2R; BDKRB2; BK-2; BK2; BK2R; BKR2; BRB2
UniProt ID: (Human) P30411, (Mouse) P32299
Entrez Gene ID: (Human) 624, (Mouse) 12062
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.