Invitrogen
Manufacturer Code:MA3082
Catalog # PIMA3082
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2SCA-1H9 |
Immunogen | Human Ataxin-3 fragment from aa F112-aa L249 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C,Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ataxin 3 variant an; ataxin 3 variant ao; ataxin 3 variant at; ataxin 3 variant e; ataxin 3 variant h; ataxin 3 variant m; ataxin 3 variant r; ataxin 3 variant ref; ataxin 3 variant y; Ataxin-3; josephin; Machado-Joseph disease (spinocerebellar ataxia 3 olivopontocerebellar ataxia 3 autosomal domit ataxin 3) Machado-Joseph disease protein 1 ataxin 3 variant h ataxin 3 variant m ataxin 3 variant ref ataxin-3 josephin olivopontocerebellar ataxia 3 spinocerebellar ataxia type 3 protein ATX3 JOS MJD MJD1 SCA3 Machado-Joseph disease (spinocerebellar ataxia 3 autosomal domit ataxin 3); Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3); Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3) homolog; Machado-Joseph disease protein 1; Machado-Joseph disease protein 1 homolog; olivopontocerebellar ataxia 3; Spinocerebellar ataxia type 3 protein
Gene Aliases: 2210008M02Rik; AI463012; AI647473; AT3; ataxin-3; ATX3; ATXN3; JOS; MJD; MJD1; Rsca3; SCA3
UniProt ID: (Human) P54252, (Rat) O35815, (Mouse) Q9CVD2
Entrez Gene ID: (Human) 4287, (Rat) 60331, (Mouse) 110616
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.