Invitrogen
Manufacturer Code:PA578848
Catalog # PIPA578848
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human Ataxin 3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ataxin 3 variant an; ataxin 3 variant ao; ataxin 3 variant at; ataxin 3 variant e; ataxin 3 variant h; ataxin 3 variant m; ataxin 3 variant r; ataxin 3 variant ref; ataxin 3 variant y; Ataxin-3; Ataxin-3 Machado-Joseph disease protein 1 Spinocerebellar ataxia type 3 protein ATXN3 ATX3 MJD MJD1 SCA3; josephin; Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3); Machado-Joseph disease protein 1; Machado-Joseph disease protein 1 homolog; olivopontocerebellar ataxia 3; Spinocerebellar ataxia type 3 protein
Gene Aliases: AT3; ATX3; ATXN3; JOS; MJD; MJD1; Rsca3; SCA3
UniProt ID: (Human) P54252, (Rat) O35815
Entrez Gene ID: (Human) 4287, (Rat) 60331
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.