Novus Biologicals
Manufacturer Code:NBP29261120UL
Catalog # NB027093
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-364 of human Ataxin 3 (NP_004984.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ataxin 3 ATX3AT3 EC 3.4.19.12 EC 3.4.22 Machado-Joseph disease protein 1 MJD1 MJDolivopontocerebellar ataxia3 autosomal dominant ataxin 3) olivopontocerebellar ataxia 3 Spinocerebellar ataxia type 3 protein; ataxin 3 variant an; ataxin 3 variant ao; ataxin 3 variant at; ataxin 3 variant e; ataxin 3 variant h; ataxin 3 variant m; ataxin 3 variant r; ataxin 3 variant ref; ataxin 3 variant y; Ataxin-3; josephin; Machado-Joseph disease (spinocerebellar ataxia 3, olivopontocerebellar ataxia 3, autosomal dominant, ataxin 3); Machado-Joseph disease protein 1; olivopontocerebellar ataxia 3; Spinocerebellar ataxia type 3 protein
Gene Aliases: AT3; ATX3; ATXN3; JOS; MJD; MJD1; SCA3
UniProt ID: (Human) P54252
Entrez Gene ID: (Human) 4287
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.