Invitrogen
Manufacturer Code:711186
Catalog # PI711186
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Oligoclonal |
Type | Antibody |
Immunogen | Protein corresponding to Human ANGPT2 (aa 250-496) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Recombinant Polyclonal Antibody
For Research Use Only
Protein Aliases: ANG-2; Angiopoietin-2; angiopoietin-2a; angiopoietin-2B; Tie2-ligand
Gene Aliases: AGPT2; Ang-2; ANG2; ANGPT2
UniProt ID: (Human) O15123, (Mouse) O35608
Entrez Gene ID: (Human) 285, (Mouse) 11601
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.