Novus Biologicals
Manufacturer Code:NBP1286410.025MG
Catalog # NB018471
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (HFHGVSNLQEFRFMIGGGC) corresponding to aa 362-380 of the C-terminal domain of Human Mouse and Rat ADPR 2 Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -80C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ACDCR2 adiponectin receptor 2 adiponectin receptor protein 2 FLJ21432 PAQR2MGC4640 Progestin and adipoQ receptor family member II; Adiponectin receptor protein 2; Progestin and adipoQ receptor family member 2; Progestin and adipoQ receptor family member II
Gene Aliases: ACDCR2; ADIPOR2; PAQR2
UniProt ID: (Human) Q86V24
Entrez Gene ID: (Human) 79602
Molecular Function: receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.