Novus Biologicals
Manufacturer Code:NBP159882
Catalog # NBP159882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC33A1(solute carrier family 33 (acetyl-CoA transporter) member 1) The peptide sequence was selected from the middle region of SLC33A1. Peptide sequence CNSVGQTAGYFLGNVLFLALESADFCNKYLRFQPQPRGIVTLSDFLFF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ACATNAcetyl-CoA transporter 1 acetyl-Coenzyme A transporter AT-1acetyl-coenzyme A transporter 1 AT1SPG42 solute carrier family 33 (acetyl-CoA transporter) member 1 Solute carrier family 33 member 1 spastic paraplegia 42 (autosomal dominant); Acetyl-coenzyme A transporter 1; AT-1; solute carrier family 33 (acetyl-CoA transporter), member 1; Solute carrier family 33 member 1
Gene Aliases: ACATN; AT-1; AT1; CCHLND; SLC33A1; SPG42
UniProt ID: (Human) O00400
Entrez Gene ID: (Human) 9197
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.