Abnova
Manufacturer Code:MAB12352
Catalog # 89-933-601
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1364CT291.108.155 |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to human AURKA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of human AURKA.
For Research Use Only
Protein Aliases: AIK; ARK-1; ARK1; AURA; Aurora 2; Aurora kinase A; aurora/IPL1-like kinase; Aurora/IPL1-related kinase 1; AURORA2; Breast tumor-amplified kinase; BTAK; MGC34538; protein phosphatase 1, regulatory subunit 47; serine/threonine protein kinase 15; Serine/threonine-protein kinase 15; Serine/threonine-protein kinase 6; Serine/threonine-protein kinase aurora-A; STK15; STK6; STK7
Gene Aliases: AIK; AIRK1; ARK1; AURA; AURKA; AYK1; BTAK; IAK1; PPP1R47; STK15; STK6; STK7
UniProt ID: (Human) O14965
Entrez Gene ID: (Human) 6790
Molecular Function: kinase non-receptor serine/threonine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.